Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOX1
Entrez Gene ID
219736
Gene Name
storkhead box 1
Gene Aliases
C10orf24
Chromosome Location
Chr.10: 68827537 - 68895942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130160.2 NP_001123632.1 2 - 3 554 123
GenBank mRNA AY842016.1 - 2 - 3 467 123

View Full Product Details