Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC13
Entrez Gene ID
6396
Gene Name
SEC13 homolog, nuclear pore and COPII coat complex component
Gene Aliases
D3S1231E, SEC13L1, SEC13R, npp-20
Chromosome Location
Chr.3: 10300929 - 10321188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136026.2 NP_001129498.1 6 - 7 765 92
NM_001136232.2 NP_001129704.1 5 - 6 619 92
NM_030673.3 NP_109598.2 6 - 7 621 92
NM_183352.2 NP_899195.1 5 - 6 587 92
XM_005265379.2 XP_005265436.1 6 - 7 638 92
XM_017007019.1 XP_016862508.1 6 - 7 675 92
XM_017007020.1 XP_016862509.1 5 - 6 482 92
XM_017007021.1 XP_016862510.1 5 - 6 492 92
GenBank mRNA AB209554.1 - 4 - 5 434 92
AF052155.1 - 5 - 6 619 92
AK095629.1 - 3 - 4 2446 92
AK223019.1 - 5 - 6 472 92
AK301999.1 - 6 - 7 689 92
BC002634.2 - 6 - 7 502 92
BC006167.1 - 2 - 3 279 92
BC091506.1 - 5 - 6 497 92
CN344033.1 - 5 - 6 599 92
EF036494.1 - 5 - 6 486 92
L09260.1 - 5 - 6 464 92

View Full Product Details