The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002483.6 | NP_002474.4 | 5 - 6 | 1198 | 125 |
GenBank mRNA | BC005008.1 | - | 5 - 6 | 1119 | 125 |
M18216.1 | - | 5 - 6 | 1125 | 124 | |
M18728.1 | - | 5 - 6 | 1129 | 125 |