The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080615.1 | NP_542182.1 | 5 - 6 | 1678 | 62 |
GenBank mRNA | AK131203.1 | - | 5 - 6 | 1678 | 62 |