Product Details

Assay Reference Genome
Location

Chr.7:44176879 on build GRCh38
Cytoband
7p13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2645
Gene Symbol
GCK
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
Location
Chr.7:44143213-44189439 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GCK NM_000162.3 NP_000153.1
XM_017011966.1 XP_016867455.1
BC001890.1 AAH01890.1
BT007062.1 AAP35711.1
EU008605.1
M88011.1 AAA51824.1
M90299.1 AAA52562.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830983 Chr7:44068842 - 44201624 on Build GRCh38 Loss POLM MYL7 MIR6838 GCK MIR4649 YKT6 POLD2 AEBP1
nsv8082 Chr7:43956156 - 44207229 on Build GRCh38 Gain MYL7 MIR6838 MIR4649 POLR2J4 YKT6 LINC00957 POLD2 AEBP1 MIR6837 DBNL POLM GCK SPDYE1 PGAM2 RASA4CP
nsv509195 Chr7:44147118 - 44261782 on Build GRCh38 Insertion GCK YKT6 CAMK2B

View Full Product Details