The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024046.1 | 2 - 3 | 241 | 64 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | neurobeachin like 2 | NBEAL2 | BDPLT4, GPS | -2.6kb | Sense | Chr. - 46979683 - 47009704 on Build GRCh38 |
Downstream | SET domain containing 2 | SETD2 | HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP | 3.6kb | Anti-sense | Chr. - 47016408 - 47164109 on Build GRCh38 |