Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADGRE4P
- Entrez Gene ID
-
326342
- Gene Name
- adhesion G protein-coupled receptor E4, pseudogene
- Gene Aliases
-
EMR4, EMR4P, FIRE, GPR127, PGR16
- Chromosome Location
-
Chr.19: 6952500 - 6990846 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024075.1 |
|
7 - 8 |
699 |
92 |
GenBank mRNA |
AF489700.1 |
- |
7 - 8 |
699 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
adhesion G protein-coupled receptor E1 |
ADGRE1 |
EMR1, TM7LN3 |
-34.0kb
|
Anti-sense |
Chr. - 6887549 - 6940453 on Build GRCh38 |
Downstream |
microtubule affinity regulating kinase 1 pseudogene |
FLJ25758 |
|
27.6kb
|
Sense |
Chr. - 7004953 - 7006415 on Build GRCh38 |
View Full Product Details