The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024092.1 | 1 - 1 | 153 | 82 | |
GenBank mRNA | AF086441.1 | - | 1 - 1 | 153 | 82 |