Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
Chromosome Location
Chr.1: 216503246 - 217137757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243515.1 NP_001230444.1 1 - 2 225 94
XM_011509267.1 XP_011507569.1 1 - 2 182 94
XM_011509276.1 XP_011507578.1 1 - 2 218 94
XM_011509279.1 XP_011507581.1 1 - 2 205 94
XM_017000628.1 XP_016856117.1 1 - 2 182 94
XM_017000630.1 XP_016856119.1 1 - 2 182 94
XM_017000635.1 XP_016856124.1 1 - 2 182 94
XM_017000641.1 XP_016856130.1 1 - 2 205 94
GenBank mRNA AK290945.1 - 1 - 2 206 94
DA722064.1 - 1 - 2 181 94


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA099848.1 - 1 - 2 205 Chr. 1 - 216677312 - 216723452 [-] on Build GRCh38 94
GenBankmRNA DA715125.1 - 1 - 2 206 Chr. 1 - 216677451 - 216723453 [-] on Build GRCh38 94
GenBankmRNA DA407868.1 - 1 - 2 182 Chr. 1 - 216707420 - 216723429 [-] on Build GRCh38 94
GenBankmRNA DA718201.1 - 1 - 2 220 Chr. 1 - 216677452 - 216723465 [-] on Build GRCh38 94


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream usherin USH2A RP39, US2, USH2, dJ1111A8.1 -291.7kb Sense Chr. - 215622894 - 216423396 on Build GRCh38
Downstream G-patch domain containing 2 GPATCH2 CT110, GPATC2, PPP1R30 703.5kb Sense Chr. - 217426992 - 217631312 on Build GRCh38

View Full Product Details