Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRKXP1
- Entrez Gene ID
-
441733
- Gene Name
- protein kinase, X-linked, pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 100547752 - 100559283 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073405.1 |
|
- |
11376 |
102 |
GenBank mRNA |
AK095793.1 |
- |
1 - 1 |
2252 |
103 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR739669.1 |
- |
1 |
|
|
Chr. 15 - 100547759 - 100548159 [+] on Build GRCh38 |
102 |
GenBankmRNA |
AI983986.1 |
- |
1 |
|
|
Chr. 15 - 100547760 - 100548237 [+] on Build GRCh38 |
101 |
GenBankmRNA |
CF887624.1 |
- |
1 |
98 |
|
Chr. 15 - 100547759 - 100548005 [-] on Build GRCh38 |
102 |
GenBankmRNA |
AI439088.1 |
- |
1 |
|
|
Chr. 15 - 100547766 - 100548165 [+] on Build GRCh38 |
102 |
GenBankmRNA |
W31675.1 |
- |
|
|
|
Chr. - - [-] on Build |
102 |
GenBankmRNA |
CF891602.1 |
- |
1 |
|
|
Chr. 15 - 100547759 - 100547989 [+] on Build GRCh38 |
102 |
GenBankmRNA |
N39117.1 |
- |
1 |
|
|
Chr. 15 - 100547761 - 100548163 [+] on Build GRCh38 |
102 |
GenBankmRNA |
DB317623.1 |
- |
1 |
|
|
Chr. 15 - 100547767 - 100548312 [+] on Build GRCh38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ceramide synthase 3 |
CERS3 |
ARCI9, LASS3 |
-3.0kb
|
Sense |
Chr. - 100400395 - 100544982 on Build GRCh38 |
Downstream |
uncharacterized LOC102723335 |
LOC102723335 |
|
8.5kb
|
Anti-sense |
Chr. - 100558666 - 100559799 on Build GRCh38 |
View Full Product Details