Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BTG3
- Entrez Gene ID
-
10950
- Gene Name
- BTG anti-proliferation factor 3
- Gene Aliases
-
ANA, APRO4, TOB5, TOB55, TOFA
- Chromosome Location
-
Chr.21: 17593650 - 17612950 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC028229.1 |
- |
1 - 1 |
1857 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV720896.1 |
- |
1 |
|
|
Chr. 21 - 17613480 - 17614051 [-] on Build GRCh38 |
90 |
GenBankmRNA |
BX101397.1 |
- |
1 |
275 |
|
Chr. 21 - 17613355 - 17613859 [+] on Build GRCh38 |
90 |
GenBankmRNA |
BU674227.1 |
- |
1 |
|
|
Chr. 21 - 17613435 - 17614114 [-] on Build GRCh38 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1549 |
LINC01549 |
C21orf37 |
-162.6kb
|
Sense |
Chr. - 17438890 - 17449185 on Build GRCh38 |
Downstream |
C21orf91 overlapping transcript 1 |
C21orf91-OT1 |
D21S2089E, NCRNA00285 |
163.3kb
|
Anti-sense |
Chr. - 17777404 - 17792509 on Build GRCh38 |
View Full Product Details