Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NRG1
- Entrez Gene ID
-
3084
- Gene Name
- neuregulin 1
- Gene Aliases
-
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
- Chromosome Location
-
Chr.8: 31639222 - 32771716 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF217878.1 |
- |
1 |
426 |
|
Chr. 8 - 32766749 - 32767452 [+] on Build GRCh38 |
79 |
GenBankmRNA |
BM553545.1 |
- |
|
214 |
|
Chr. - - [-] on Build |
79 |
GenBankmRNA |
CD676708.1 |
- |
1 |
230 |
|
Chr. 8 - 32766946 - 32767557 [+] on Build GRCh38 |
79 |
GenBankmRNA |
EL951449.1 |
- |
1 |
166 |
|
Chr. 8 - 32767039 - 32767607 [+] on Build GRCh38 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NRG1 intronic transcript 3 |
NRG1-IT3 |
|
-323.4kb
|
Sense |
Chr. - 32440746 - 32442725 on Build GRCh38 |
Downstream |
uncharacterized LOC105379362 |
LOC105379362 |
|
160.1kb
|
Sense |
Chr. - 32927942 - 33050201 on Build GRCh38 |
View Full Product Details