Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DRD3
- Entrez Gene ID
-
1814
- Gene Name
- dopamine receptor D3
- Gene Aliases
-
D3DR, ETM1, FET1
- Chromosome Location
-
Chr.3: 114127623 - 114199407 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY228459.1 |
- |
1 - 1 |
64 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
queuine tRNA-ribosyltransferase accessory subunit 2 |
QTRT2 |
QTRTD1 |
-83.6kb
|
Anti-sense |
Chr. - 114056735 - 114088422 on Build GRCh38 |
Downstream |
zinc finger protein 80 |
ZNF80 |
pT17 |
62.5kb
|
Sense |
Chr. - 114211035 - 114237578 on Build GRCh38 |
View Full Product Details