Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EIF4A2
- Entrez Gene ID
-
1974
- Gene Name
- eukaryotic translation initiation factor 4A2
- Gene Aliases
-
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
- Chromosome Location
-
Chr.3: 186783572 - 186789897 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC039344.1 |
- |
1 - 1 |
1956 |
79 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX118227.1 |
- |
1 |
414 |
|
Chr. 3 - 186781904 - 186782641 [-] on Build GRCh38 |
79 |
GenBankmRNA |
HY217426.1 |
- |
1 |
|
|
Chr. 3 - 186781786 - 186782303 [+] on Build GRCh38 |
79 |
GenBankmRNA |
AA496083.1 |
- |
1 |
414 |
|
Chr. 3 - 186782152 - 186782641 [-] on Build GRCh38 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
kininogen 1 |
KNG1 |
BDK, BK, KNG |
-37.4kb
|
Anti-sense |
Chr. - 186717309 - 186744410 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 2 |
SNORD2 |
R39B, SNR39B |
617.0
|
Anti-sense |
Chr. - 186784796 - 186784865 on Build GRCh38 |
View Full Product Details