Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FUT4
- Entrez Gene ID
-
2526
- Gene Name
- fucosyltransferase 4
- Gene Aliases
-
CD15, ELFT, FCT3A, FUC-TIV, FUTIV, LeX, SSEA-1
- Chromosome Location
-
Chr.11: 94543851 - 94549898 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC041922.1 |
- |
1 - 1 |
1754 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI796025.1 |
- |
1 |
|
|
Chr. 11 - 94542961 - 94543372 [+] on Build GRCh38 |
69 |
GenBankmRNA |
BM930245.1 |
- |
1 |
396 |
|
Chr. 11 - 94542957 - 94543424 [-] on Build GRCh38 |
69 |
GenBankmRNA |
DB549341.1 |
- |
1 |
393 |
|
Chr. 11 - 94542964 - 94543421 [-] on Build GRCh38 |
69 |
GenBankmRNA |
BM930393.1 |
- |
1 |
396 |
|
Chr. 11 - 94542960 - 94543424 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 11 open reading frame 97 |
C11orf97 |
LINC01171 |
-10.8kb
|
Anti-sense |
Chr. - 94512529 - 94532123 on Build GRCh38 |
Downstream |
piwi like RNA-mediated gene silencing 4 |
PIWIL4 |
HIWI2, MIWI2 |
22.5kb
|
Anti-sense |
Chr. - 94567308 - 94621421 on Build GRCh38 |
View Full Product Details