Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LRRC8B
- Entrez Gene ID
-
23507
- Gene Name
- leucine rich repeat containing 8 family member B
- Gene Aliases
-
TA-LRRP, TALRRP
- Chromosome Location
-
Chr.1: 89522627 - 89597861 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC053565.1 |
- |
1 - 1 |
51 |
74 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ717287.1 |
- |
1 |
50 |
|
Chr. 1 - 89584150 - 89584917 [-] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
guanylate binding protein 1 pseudogene 1 |
GBP1P1 |
|
-157.7kb
|
Anti-sense |
Chr. - 89407679 - 89424934 on Build GRCh38 |
Downstream |
leucine rich repeat containing 8 family member C |
LRRC8C |
AD158, FAD158 |
31.0kb
|
Anti-sense |
Chr. - 89615810 - 89719535 on Build GRCh38 |
View Full Product Details