Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CELF3
- Entrez Gene ID
-
11189
- Gene Name
- CUGBP, Elav-like family member 3
- Gene Aliases
-
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
- Chromosome Location
-
Chr.1: 151700058 - 151716821 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP394901.1 |
- |
1 |
359 |
|
Chr. 1 - 151700058 - 151700639 [-] on Build GRCh38 |
100 |
GenBankmRNA |
DB294995.1 |
- |
1 |
|
|
Chr. 1 - 151700060 - 151700465 [+] on Build GRCh38 |
100 |
GenBankmRNA |
DB301268.1 |
- |
1 |
|
|
Chr. 1 - 151700060 - 151700581 [+] on Build GRCh38 |
100 |
GenBankmRNA |
BM676034.1 |
- |
1 |
|
|
Chr. 1 - 151700058 - 151700655 [+] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sorting nexin family member 27 |
SNX27 |
MRT1, MY014 |
-977.0
|
Anti-sense |
Chr. - 151612027 - 151699083 on Build GRCh38 |
Downstream |
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1 |
RIIAD1 |
C1orf230, NCRNA00166 |
19.1kb
|
Anti-sense |
Chr. - 151721537 - 151729606 on Build GRCh38 |
View Full Product Details