Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- XPO4
- Entrez Gene ID
-
64328
- Gene Name
- exportin 4
- Gene Aliases
-
exp4
- Chromosome Location
-
Chr.13: 20777329 - 20902774 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK129848.1 |
- |
1 - 1 |
1541 |
74 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
T93310.1 |
- |
|
|
|
Chr. - - [-] on Build |
74 |
GenBankmRNA |
AA737176.1 |
- |
1 |
|
|
Chr. 13 - 20831810 - 20832139 [+] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 |
EEF1AKMT1 |
ESP13, N6AMT2 |
-57.9kb
|
Sense |
Chr. - 20728931 - 20773958 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 367 |
LINC00367 |
|
105.1kb
|
Sense |
Chr. - 20938564 - 20949428 on Build GRCh38 |
View Full Product Details