Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RASAL2
- Entrez Gene ID
-
9462
- Gene Name
- RAS protein activator like 2
- Gene Aliases
-
NGAP
- Chromosome Location
-
Chr.1: 178093729 - 178479514 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK127614.1 |
- |
1 - 1 |
951 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RASAL2 antisense RNA 1 |
RASAL2-AS1 |
|
-38.4kb
|
Anti-sense |
Chr. - 178091508 - 178093993 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 83 |
LINC00083 |
NCRNA00083 |
343.7kb
|
Anti-sense |
Chr. - 178479514 - 178499623 on Build GRCh38 |
View Full Product Details