Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
Chromosome Location
Chr.9: 89318473 - 89367117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK127655.1 - 1 - 1 2352 64


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF433336.1 - 1 Chr. 9 - 89355072 - 89355591 [-] on Build GRCh38 64
GenBankmRNA DB282642.1 - 1 488 Chr. 9 - 89354986 - 89355536 [+] on Build GRCh38 64
GenBankmRNA DB321412.1 - 1 Chr. 9 - 89355059 - 89355590 [-] on Build GRCh38 64
GenBankmRNA AI498974.1 - 1 Chr. 9 - 89355097 - 89355590 [-] on Build GRCh38 64
GenBankmRNA DA222403.1 - 1 72 Chr. 9 - 89355402 - 89356054 [+] on Build GRCh38 64
GenBankmRNA BE674458.1 - 1 Chr. 9 - 89355142 - 89355593 [-] on Build GRCh38 64


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream CDC28 protein kinase regulatory subunit 2 CKS2 CKSHS2 -36.4kb Sense Chr. - 89311195 - 89316703 on Build GRCh38
Downstream semaphorin 4D SEMA4D C9orf164, CD100, M-sema-G, SEMAJ, coll-4 5.2kb Anti-sense Chr. - 89360791 - 89498065 on Build GRCh38

View Full Product Details