Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100130913
- Entrez Gene ID
-
100130913
- Gene Name
- uncharacterized LOC100130913
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK128355.1 |
- |
1 - 1 |
2751 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB346209.1 |
- |
1 |
|
|
Chr. 7 - 55442108 - 55442652 [-] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
vesicular, overexpressed in cancer, prosurvival protein 1 |
VOPP1 |
ECOP, GASP |
|
Anti-sense |
Chr. - 55434406 - 55572520 on Build GRCh38 |
Downstream |
FK506 binding protein 9 pseudogene 1 |
FKBP9P1 |
FKBP9L |
238.4kb
|
Anti-sense |
Chr. - 55681074 - 55704567 on Build GRCh38 |
View Full Product Details