Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC070492.1 - 8 - 9 762 Chr. 15 - 32398950 - 32435233 [-] on Build GRCh38 112


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream golgin A8 family member K GOLGA8K Sense Chr. - 32389585 - 32403292 on Build GRCh38
Downstream ULK4 pseudogene 1 ULK4P1 D-X, FAM7A1 313.0 Sense Chr. - 32406611 - 32435049 on Build GRCh38

View Full Product Details