Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KDM4C
- Entrez Gene ID
-
23081
- Gene Name
- lysine demethylase 4C
- Gene Aliases
-
GASC1, JHDM3C, JMJD2C, TDRD14C
- Chromosome Location
-
Chr.9: 6720863 - 7175648 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF085959.1 |
- |
1 - 1 |
550 |
68 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA019195.1 |
- |
1 |
|
|
Chr. 9 - 6909181 - 6909614 [-] on Build GRCh38 |
68 |
GenBankmRNA |
BX115579.1 |
- |
1 |
550 |
|
Chr. 9 - 6908974 - 6909628 [+] on Build GRCh38 |
68 |
GenBankmRNA |
H68313.1 |
- |
1 |
|
|
Chr. 9 - 6909230 - 6909614 [-] on Build GRCh38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glycine decarboxylase |
GLDC |
GCE, GCSP, HYGN1 |
-263.3kb
|
Anti-sense |
Chr. - 6532464 - 6645692 on Build GRCh38 |
Downstream |
transmembrane protein 261 |
TMEM261 |
C9orf123 |
886.9kb
|
Anti-sense |
Chr. - 7796490 - 7799806 on Build GRCh38 |
View Full Product Details