Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ACAT1
- Entrez Gene ID
-
38
- Gene Name
- acetyl-CoA acetyltransferase 1
- Gene Aliases
-
ACAT, MAT, T2, THIL
- Chromosome Location
-
Chr.11: 108121531 - 108148168 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF086561.1 |
- |
1 - 1 |
549 |
85 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW750026.1 |
- |
1 |
328 |
|
Chr. 11 - 108147843 - 108148271 [+] on Build GRCh38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cullin 5 |
CUL5 |
VACM-1, VACM1 |
-39.8kb
|
Sense |
Chr. - 108008682 - 108107766 on Build GRCh38 |
Downstream |
nuclear protein, coactivator of histone transcription |
NPAT |
E14, E14/NPAT, p220 |
9.0kb
|
Anti-sense |
Chr. - 108155280 - 108222638 on Build GRCh38 |
View Full Product Details