The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024393.1 | 3 - 4 | 954 | 96 | |
NR_117100.1 | 3 - 4 | 954 | 96 | ||
GenBank mRNA | DQ515896.1 | - | 3 - 4 | 954 | 96 |
DQ515897.1 | - | 3 - 4 | 954 | 96 |