Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
Chromosome Location
Chr.18: 62715439 - 62980443 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF130097.2 - 1 - 1 677 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream zinc finger CCHC-type containing 2 ZCCHC2 C18orf49 -250.0kb Anti-sense Chr. - 62523410 - 62587709 on Build GRCh38
Downstream BCL2, apoptosis regulator BCL2 Bcl-2, PPP1R50 283.5kb Sense Chr. - 63123346 - 63319778 on Build GRCh38

View Full Product Details