Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01181
- Entrez Gene ID
-
379034
- Gene Name
- long intergenic non-protein coding RNA 1181
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001351.1 |
- |
4 - 5 |
2026 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
BAALC antisense RNA 2 |
BAALC-AS2 |
BAALCOS, C8orf56 |
|
Anti-sense |
Chr. - 103132963 - 103141342 on Build GRCh38 |
Downstream |
brain and acute leukemia, cytoplasmic |
BAALC |
|
1.3kb
|
Sense |
Chr. - 103140693 - 103230305 on Build GRCh38 |
View Full Product Details