Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DNM3OS
- Entrez Gene ID
-
100628315
- Gene Name
- DNM3 opposite strand/antisense RNA
- Gene Aliases
-
DNM3-AS1, MIR199A2HG
- Chromosome Location
-
Chr.1: 172136879 - 172144835 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038397.2 |
|
1 - 1 |
4594 |
111 |
NR_103486.1 |
|
4 - 4 |
688 |
111 |
GenBank mRNA |
AK021543.1 |
- |
1 - 1 |
348 |
111 |
FJ623959.1 |
- |
1 - 1 |
4594 |
111 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AU117617.1 |
- |
1 |
348 |
|
Chr. 1 - 172139707 - 172140589 [-] on Build GRCh38 |
111 |
GenBankmRNA |
AU155531.1 |
- |
1 |
|
|
Chr. 1 - 172140081 - 172140536 [+] on Build GRCh38 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 214 |
MIR214 |
MIRN214, miRNA214, mir-214 |
-145.0
|
Sense |
Chr. - 172138798 - 172138907 on Build GRCh38 |
Downstream |
microRNA 199a-2 |
MIR199A2 |
MIR-199-s, MIRN199A2, mir-199a-2 |
3.9kb
|
Sense |
Chr. - 172144535 - 172144644 on Build GRCh38 |
View Full Product Details