Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SCARB2
- Entrez Gene ID
-
950
- Gene Name
- scavenger receptor class B member 2
- Gene Aliases
-
AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII
- Chromosome Location
-
Chr.4: 76158737 - 76213899 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK022432.1 |
- |
1 - 1 |
1146 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nucleoporin 54 |
NUP54 |
|
-24.2kb
|
Sense |
Chr. - 76114659 - 76148515 on Build GRCh38 |
Downstream |
family with sequence similarity 47 member E |
FAM47E |
|
38.6kb
|
Anti-sense |
Chr. - 76214040 - 76283783 on Build GRCh38 |
View Full Product Details