Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SIAH1
- Entrez Gene ID
-
6477
- Gene Name
- siah E3 ubiquitin protein ligase 1
- Gene Aliases
-
SIAH1A
- Chromosome Location
-
Chr.16: 48360531 - 48448440 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK023458.1 |
- |
1 - 1 |
1257 |
91 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF989665.1 |
- |
1 |
81 |
|
Chr. 16 - 48364358 - 48364695 [-] on Build GRCh38 |
91 |
GenBankmRNA |
DA096768.1 |
- |
1 |
422 |
|
Chr. 16 - 48364477 - 48365050 [-] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100507577 |
LOC100507577 |
|
-970.0
|
Anti-sense |
Chr. - 48355685 - 48362999 on Build GRCh38 |
Downstream |
NEDD4 binding protein 1 |
N4BP1 |
|
172.8kb
|
Sense |
Chr. - 48538726 - 48610214 on Build GRCh38 |
View Full Product Details