Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCA1
Entrez Gene ID
79781
Gene Name
IQ motif containing with AAA domain 1
Gene Aliases
4930465P12Rik, DRC11, IQCA
Chromosome Location
Chr.2: 236324147 - 236507558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC008085.1 - 1 - 1 751 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ankyrin repeat and SOCS box containing 18 ASB18 ASB-18 -103.2kb Anti-sense Chr. - 236194872 - 236264345 on Build GRCh38
Downstream atypical chemokine receptor 3 ACKR3 CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 168.0kb Sense Chr. - 236537131 - 236582358 on Build GRCh38

View Full Product Details