The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AB064670.1 | - | 1 - 1 | 248 | 85 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | glycerol-3-phosphate acyltransferase 3 | GPAT3 | AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1 | Anti-sense | Chr. - 83535634 - 83605875 on Build GRCh38 | |
Downstream | uncharacterized LOC101928978 | LOC101928978 | 406.3kb | Sense | Chr. - 83968082 - 84299169 on Build GRCh38 |