The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026679.1 | 8 - 9 | 1912 | 102 | |
GenBank mRNA | AF418573.2 | - | 8 - 9 | 1912 | 102 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | TEK receptor tyrosine kinase | TEK | CD202B, TIE-2, TIE2, VMCM, VMCM1 | -41.7kb | Anti-sense | Chr. - 27109141 - 27230178 on Build GRCh38 |
Downstream | equatorin | EQTN | AFAF, C9orf11, SPACA8 | 10.4kb | Sense | Chr. - 27284654 - 27297139 on Build GRCh38 |