The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK055847.1 | - | 1 - 1 | 644 | 78 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | charged multivesicular body protein 5 | CHMP5 | C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60 | -64.4kb | Sense | Chr. - 33264879 - 33282069 on Build GRCh38 |
Downstream | aquaporin 7 | AQP7 | AQP7L, AQPap, GLYCQTL | 36.4kb | Anti-sense | Chr. - 33383135 - 33402682 on Build GRCh38 |