Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CENPL
- Entrez Gene ID
-
91687
- Gene Name
- centromere protein L
- Gene Aliases
-
C1orf155, CENP-L, dJ383J4.3
- Chromosome Location
-
Chr.1: 173799550 - 173824639 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK056348.1 |
- |
1 - 1 |
1451 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
aspartyl-tRNA synthetase 2, mitochondrial |
DARS2 |
ASPRS, LBSL, MT-ASPRS |
|
Anti-sense |
Chr. - 173824659 - 173858544 on Build GRCh38 |
Downstream |
GAS5 antisense RNA 1 |
GAS5-AS1 |
|
38.4kb
|
Anti-sense |
Chr. - 173863248 - 173863941 on Build GRCh38 |
View Full Product Details