Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARPP21
- Entrez Gene ID
-
10777
- Gene Name
- cAMP regulated phosphoprotein 21
- Gene Aliases
-
ARPP-21, R3HDM3, RCS, TARPP
- Chromosome Location
-
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK091377.1 |
- |
1 - 1 |
1844 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 128-2 |
MIR128-2 |
MIR128B, MIRN128-2, MIRN128B, mir-128-2, mir-128b |
-39.5kb
|
Sense |
Chr. - 35744476 - 35744559 on Build GRCh38 |
Downstream |
SH3 and cysteine rich domain |
STAC |
STAC1 |
593.9kb
|
Sense |
Chr. - 36380455 - 36548007 on Build GRCh38 |
View Full Product Details