Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TAF1
- Entrez Gene ID
-
6872
- Gene Name
- TATA-box binding protein associated factor 1
- Gene Aliases
-
BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP
- Chromosome Location
-
Chr.X: 71366220 - 71530525 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104396.1 |
|
1 - 1 |
967 |
73 |
GenBank mRNA |
AJ549246.2 |
- |
1 - 1 |
967 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
inhibitor of growth family, X-linked (pseudogene) |
INGX |
ING1-like, ING2 |
|
Anti-sense |
Chr. - 71491681 - 71492449 on Build GRCh38 |
Downstream |
O-linked N-acetylglucosamine (GlcNAc) transferase |
OGT |
HINCUT-1, HRNT1, O-GLCNAC |
40.1kb
|
Sense |
Chr. - 71533062 - 71575897 on Build GRCh38 |
View Full Product Details