Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TOM1L2
- Entrez Gene ID
-
146691
- Gene Name
- target of myb1 like 2 membrane trafficking protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 17843508 - 17972470 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK025154.1 |
- |
1 - 1 |
2651 |
98 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP430199.1 |
- |
1 |
169 |
|
Chr. 17 - 17865867 - 17866279 [-] on Build GRCh38 |
98 |
GenBankmRNA |
AV703573.1 |
- |
|
151 |
|
Chr. - - [-] on Build |
98 |
GenBankmRNA |
HY327671.1 |
- |
1 |
|
|
Chr. 17 - 17865763 - 17866207 [+] on Build GRCh38 |
98 |
GenBankmRNA |
DB360447.1 |
- |
1 |
|
|
Chr. 17 - 17865763 - 17866306 [+] on Build GRCh38 |
98 |
GenBankmRNA |
EB386267.1 |
- |
1 |
165 |
|
Chr. 17 - 17865666 - 17866275 [-] on Build GRCh38 |
98 |
GenBankmRNA |
HY304797.1 |
- |
1 |
|
|
Chr. 17 - 17865763 - 17866215 [+] on Build GRCh38 |
98 |
GenBankmRNA |
HY308442.1 |
- |
1 |
|
|
Chr. 17 - 17865764 - 17866253 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100507131 |
LOC100507131 |
|
-5.1kb
|
Sense |
Chr. - 17858227 - 17860803 on Build GRCh38 |
Downstream |
dynein regulatory complex subunit 3 |
DRC3 |
CFAP134, LRRC48 |
104.0kb
|
Anti-sense |
Chr. - 17972766 - 18016889 on Build GRCh38 |
View Full Product Details