Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KLHL25
- Entrez Gene ID
-
64410
- Gene Name
- kelch like family member 25
- Gene Aliases
-
ENC-2, ENC2
- Chromosome Location
-
Chr.15: 85759326 - 85794958 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK126962.1 |
- |
2 - 2 |
3456 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101929679 |
LOC101929679 |
|
-605.0
|
Sense |
Chr. - 85754941 - 85758725 on Build GRCh38 |
Downstream |
microRNA 1276 |
MIR1276 |
MIRN1276, hsa-mir-1276, mir-1276 |
8.1kb
|
Sense |
Chr. - 85770496 - 85770578 on Build GRCh38 |
View Full Product Details