Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C2orf72
- Entrez Gene ID
-
257407
- Gene Name
- chromosome 2 open reading frame 72
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 231037566 - 231049714 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF086098.1 |
- |
1 - 1 |
449 |
77 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
N62206.1 |
- |
1 |
|
|
Chr. 2 - 231052188 - 231052632 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AA100715.1 |
- |
1 |
|
|
Chr. 2 - 231052098 - 231052635 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
proteasome 26S subunit, non-ATPase 1 |
PSMD1 |
P112, Rpn2, S1 |
4.2kb
|
Sense |
Chr. - 231056864 - 231172827 on Build GRCh38 |
View Full Product Details