Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDE4DIP
- Entrez Gene ID
-
9659
- Gene Name
- phosphodiesterase 4D interacting protein
- Gene Aliases
-
CMYA2, MMGL
- Chromosome Location
-
Chr.1: 148808167 - 149051277 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124487.1 |
- |
1 - 1 |
1747 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ188556.1 |
- |
1 |
144 |
|
Chr. 1 - 148812198 - 148812895 [+] on Build GRCh38 |
90 |
GenBankmRNA |
BI554061.1 |
- |
1 |
156 |
|
Chr. 1 - 148812186 - 148813076 [+] on Build GRCh38 |
90 |
GenBankmRNA |
CR738692.1 |
- |
|
277 |
|
Chr. - - [-] on Build |
90 |
GenBankmRNA |
HY352461.1 |
- |
1 |
|
|
Chr. 1 - 120697279 - 120697740 [+] on Build GRCh38 |
90 |
GenBankmRNA |
AA410206.1 |
- |
|
276 |
|
Chr. - - [-] on Build |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
vesicle-trafficking protein SEC22b |
LOC100996517 |
|
-18.8kb
|
Anti-sense |
Chr. - 148766340 - 148786183 on Build GRCh38 |
Downstream |
neuroblastoma breakpoint family member 9 |
NBPF9 |
AE01 |
241.0kb
|
Anti-sense |
Chr. - 149054033 - 149090861 on Build GRCh38 |
View Full Product Details