Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL36AP33
- Entrez Gene ID
-
100191039
- Gene Name
- ribosomal protein L36a pseudogene 33
- Gene Aliases
-
RPL36A_15_975
- Chromosome Location
-
Chr.9: 35507534 - 35507931 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC020828.1 |
- |
1 - 1 |
48 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG623215.1 |
- |
1 |
48 |
|
Chr. 9 - 35507479 - 35508164 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATPase phospholipid transporting 8B5, pseudogene |
ATP8B5P |
FetA |
-24.5kb
|
Sense |
Chr. - 35406755 - 35483029 on Build GRCh38 |
Downstream |
family with sequence similarity 166 member B |
FAM166B |
|
53.1kb
|
Anti-sense |
Chr. - 35561830 - 35563899 on Build GRCh38 |
View Full Product Details