Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WIPF2
Entrez Gene ID
147179
Gene Name
WAS/WASL interacting protein family member 2
Gene Aliases
WICH, WIRE
Chromosome Location
Chr.17: 40219304 - 40284136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005257083.2 XP_005257140.1 8 - 8 7057 106
XM_005257084.2 XP_005257141.1 8 - 8 7015 106
XM_011524412.1 XP_011522714.1 7 - 7 6956 106
XM_011524413.1 XP_011522715.1 7 - 7 6937 106
GenBank mRNA BC030561.1 - 1 - 1 1050 106


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI939515.1 - 1 Chr. 17 - 40283586 - 40284130 [-] on Build GRCh38 106
GenBankmRNA BE217997.1 - 1 Chr. 17 - 40283618 - 40284132 [-] on Build GRCh38 106
GenBankmRNA CA423113.1 - 1 Chr. 17 - 40282819 - 40284131 [-] on Build GRCh38 106
GenBankmRNA CA447477.1 - 1 Chr. 17 - 40280048 - 40284131 [-] on Build GRCh38 106
GenBankmRNA AI683123.1 - 1 Chr. 17 - 40283593 - 40284131 [-] on Build GRCh38 106
GenBankmRNA CA423189.1 - 1 Chr. 17 - 40282861 - 40284131 [-] on Build GRCh38 106
GenBankmRNA AW295547.1 - 1 Chr. 17 - 40283577 - 40284136 [-] on Build GRCh38 106


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Rap guanine nucleotide exchange factor like 1 RAPGEFL1 Link-GEFII -87.0kb Sense Chr. - 40177010 - 40195656 on Build GRCh38
Downstream cell division cycle 6 CDC6 CDC18L, HsCDC18, HsCDC6 3.6kb Sense Chr. - 40287689 - 40303161 on Build GRCh38

View Full Product Details