Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBE2G2
- Entrez Gene ID
-
7327
- Gene Name
- ubiquitin conjugating enzyme E2 G2
- Gene Aliases
-
UBC7
- Chromosome Location
-
Chr.21: 44768580 - 44801836 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC054341.1 |
- |
1 - 1 |
453 |
87 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
EL584630.1 |
- |
|
373 |
|
Chr. - - [-] on Build |
87 |
GenBankmRNA |
DN912953.1 |
- |
1 |
62 |
|
Chr. 21 - 44800868 - 44801406 [-] on Build GRCh38 |
87 |
GenBankmRNA |
EL585000.1 |
- |
1 |
|
|
Chr. 21 - 44800959 - 44801481 [+] on Build GRCh38 |
87 |
GenBankmRNA |
BQ424505.1 |
- |
1 |
461 |
|
Chr. 21 - 44801004 - 44801794 [-] on Build GRCh38 |
87 |
GenBankmRNA |
EL584636.1 |
- |
1 |
163 |
|
Chr. 21 - 44800959 - 44801479 [-] on Build GRCh38 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
thrombospondin type laminin G domain and EAR repeats |
TSPEAR |
C21orf29, DFNB98, TSP-EAR |
-88.4kb
|
Sense |
Chr. - 44497892 - 44711580 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1424 |
LINC01424 |
|
444.0
|
Anti-sense |
Chr. - 44802241 - 44804717 on Build GRCh38 |
View Full Product Details