Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATP2B4
- Entrez Gene ID
-
493
- Gene Name
- ATPase plasma membrane Ca2+ transporting 4
- Gene Aliases
-
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
- Chromosome Location
-
Chr.1: 203626787 - 203744081 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC062445.1 |
- |
1 - 1 |
844 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU172356.1 |
- |
1 |
487 |
|
Chr. 1 - 203733342 - 203734153 [+] on Build GRCh38 |
92 |
GenBankmRNA |
BE742459.1 |
- |
1 |
250 |
|
Chr. 1 - 203733575 - 203734319 [+] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 260 |
LINC00260 |
C1orf217, NCRNA00260 |
-1.1kb
|
Sense |
Chr. - 203730577 - 203731853 on Build GRCh38 |
Downstream |
lymphocyte transmembrane adaptor 1 |
LAX1 |
LAX |
31.0kb
|
Sense |
Chr. - 203765132 - 203778175 on Build GRCh38 |
View Full Product Details