Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PAX8
- Entrez Gene ID
-
7849
- Gene Name
- paired box 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 113215997 - 113278950 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR936798.1 |
- |
1 - 1 |
5948 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
pleckstrin and Sec7 domain containing 4 |
PSD4 |
EFA6B, TIC |
-14.1kb
|
Anti-sense |
Chr. - 113157399 - 113203229 on Build GRCh38 |
Downstream |
PAX8 antisense RNA 1 |
PAX8-AS1 |
|
10.0kb
|
Anti-sense |
Chr. - 113235527 - 113267023 on Build GRCh38 |
View Full Product Details