Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DET1
- Entrez Gene ID
-
55070
- Gene Name
- de-etiolated homolog 1 (Arabidopsis)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 88501924 - 88546703 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026645.2 |
|
3 - 4 |
1122 |
79 |
GenBank mRNA |
BC064591.1 |
- |
3 - 4 |
1079 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
mitochondrial ribosomal protein S11 |
MRPS11 |
HCC-2, MRP-S11, S11mt |
-38.2kb
|
Anti-sense |
Chr. - 88467453 - 88480767 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1586 |
LINC01586 |
|
54.1kb
|
Sense |
Chr. - 88585571 - 88605110 on Build GRCh38 |
View Full Product Details