Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BPHL
- Entrez Gene ID
-
670
- Gene Name
- biphenyl hydrolase-like (serine hydrolase)
- Gene Aliases
-
BPH-RP, MCNAA, VACVASE
- Chromosome Location
-
Chr.6: 3118376 - 3153198 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG714668.1 |
- |
1 - 2 |
473 |
|
Chr. 6 - 3118376 - 3119503 [+] on Build GRCh38 |
96 |
GenBankmRNA |
DA143212.1 |
- |
1 - 2 |
122 |
|
Chr. 6 - 3118722 - 3127348 [+] on Build GRCh38 |
96 |
GenBankmRNA |
DB466566.1 |
- |
1 - 2 |
114 |
|
Chr. 6 - 3118730 - 3123755 [+] on Build GRCh38 |
96 |
GenBankmRNA |
DB482292.1 |
- |
1 - 2 |
114 |
|
Chr. 6 - 3118730 - 3127292 [+] on Build GRCh38 |
96 |
GenBankmRNA |
DA823359.1 |
- |
1 - 2 |
118 |
|
Chr. 6 - 3118726 - 3127352 [+] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
receptor interacting serine/threonine kinase 1 |
RIPK1 |
RIP, RIP-1, RIP1 |
-3.2kb
|
Sense |
Chr. - 3063888 - 3115187 on Build GRCh38 |
Downstream |
tubulin beta 2A class IIa |
TUBB2A |
CDCBM5, TUBB, TUBB2 |
34.2kb
|
Anti-sense |
Chr. - 3153666 - 3157549 on Build GRCh38 |
View Full Product Details