Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS7
Entrez Gene ID
344805
Gene Name
transmembrane protease, serine 7
Gene Aliases
-
Chromosome Location
Chr.3: 112039067 - 112081269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026734.1 4 - 5 522 66
GenBank mRNA AK131211.1 - 4 - 5 522 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA744077.1 - 4 - 5 522 Chr. 3 - 112039618 - 112047456 [+] on Build GRCh38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transgelin 3 TAGLN3 NP22, NP24, NP25 -31.9kb Sense Chr. - 111998739 - 112013888 on Build GRCh38
Downstream chromosome 3 open reading frame 52 C3orf52 TTMP 38.8kb Sense Chr. - 112086335 - 112131095 on Build GRCh38

View Full Product Details