The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026736.1 | 2 - 3 | 292 | 62 | |
GenBank mRNA | AI621292.1 | - | 4 - 5 | 62 | |
BX280692.1 | - | 2 - 3 | 292 | 62 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group) | GCNT2 | CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1 | -55.5kb | Anti-sense | Chr. - 10521283 - 10629368 on Build GRCh38 |
Downstream | PAK1 interacting protein 1 | PAK1IP1 | MAK11, PIP1, WDR84, bA421M1.5, hPIP1 | 3.7kb | Anti-sense | Chr. - 10690865 - 10709782 on Build GRCh38 |